missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Lightning-Link (R) APC-Cy5.5 Antibody Labeling Kit
Click to view available options
Quantity:
1 mg
100 μg
30 μg
300 μg
Unit Size:
1 set
100µg
1mg
Description
The antibody is covalently bonded to the label in a directional and controlled process at near-neutral pH. APC/Cy5.5 is a tandem conjugate. In the Lightning-Link APC/Cy5.5 the tandem is excited at 633nm, wavelength at which the APC has its maximum absorption. The excited APC functions then as an energy donor, where the energy is transferred via FRET (Fluorescence Resonance Energy Transfer) to the Cy5.5. The Cy5.5 emits then the energy received from the APC in the form of long wavelength light at 694nm. The amount of antibody used for labeling should be the same as the pack size of LL-APC-Cy5.5. For any new antibody, optimization of the ratio of antibody to dye is often worthwhile. If your antibody needs to be concentrated, purified, or cleaned up, consider purchasing an antibody clean up kit.
Order Info
Quantity

Specifications
Specifications
Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
Quantity | 30 μg |
Conjugate | APC/Cy5.5 |
Kit Contents | Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL Modifier reagent, 1 vial of LL Quencher FD reagent |
Product Type | APC-Cy5.5 Antibody Labeling Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Novus Biologicals™ Lightning-Link (R) APC-Cy5.5 Antibody Labeling Kit >
Spot an opportunity for improvement?Share a Content Correction