missing translation for 'onlineSavingsMsg'
Learn More
Novus Biologicals™ Lightning-Link (R) APC-Cy7 Antibody Labeling Kit Product Code.: 18731204

Novus Biologicals™ Lightning-Link (R) APC-Cy7 Antibody Labeling Kit

Product Code. 18731204 Shop All Bio Techne Products
1 mg, 1mg
Click to view available options
Quantity:
1 mg
1 x 100 ÎĽg Reaction
3 x 0.01 mg Reaction
300 ÎĽg
Unit Size:
1 set
100µg
1mg
30µg
This item is not returnable. View return policy

Product Code. 18731204

Brand: Novus Biologicals™ 7650015

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Applications: Flow Cytometry

APC/Cy7 is a tandem conjugate. In the Lightning-Link APC/Cy7 the tandem is excited at 633nm, wavelength at which the APC has its maximum absorption. The excited APC functions then as an energy donor, where the energy is transferred via FRET (Fluorescence Resonance Energy Transfer) to the Cy7. The Cy7 emits then the energy received from the APC in the form of long wavelength light at 776nm.

Order Info

Quantity

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Quantity 1 mg
Conjugate APC/Cy7
Kit Contents Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL Modifier reagent, 1 vial of LL Quencher FD reagent
Product Type APC-Cy7 Antibody Labeling Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Novus Biologicals™ Lightning-Link (R) APC-Cy7 Antibody Labeling Kit >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.