missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIN37 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | LIN37 |
---|---|
Concentration | 0.33 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16885055
|
Proteintech
26651-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16875055
|
Proteintech
26651-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LIN37, also named as Antolefinin and MSTP064, is a component of the DREAM complex (also named LINC complex). LIN37 migrates to 37 kDa for modification.Specifications
LIN37 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q96GY3 | |
55957 | |
LIN37 Fusion Protein Ag24565 | |
Primary | |
-20°C | |
LIN37 |
0.33 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Antolefinin, F25965, lin 37, lin 37 homolog (C. elegans), LIN37, Protein lin 37 homolog, ZK418.4 | |
LIN37 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
LIN37 Rabbit anti-Human, Polyclonal, Proteintech