missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium Nitrate Crystals, Ultra-Pure Additive, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
2110.00 SEK - 4346.00 SEK
Specifications
Grade | Ultrapure |
---|---|
Chemical Name or Material | Lithium Nitrate Crystals, Additive Ultra Pure |
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
- Ultra Pure Additives
- Lithium Nitrate Crystals
- 500 g Package Size
Certifications
ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015
Compliance
GHS/OSHA Complaint
Specifications
Ultrapure | |
Lithium Nitrate Crystals, Additive Ultra Pure |
Safety and Handling
missing translation for 'dotInformation' : Nonhazardous
Recommended Storage : Ambient
Product intended for laboratory use only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Lithium Nitrate Crystals, Ultra-Pure Additive, SPEX CertiPrep™