Learn More
Litmus Solution, Pure, Indicator Grade, Fisher Chemical™ Product Code.: 10284340
Litmus Solution, Pure, Indicator Grade, Fisher Chemical™
Product Code. p-7073271 Shop All Fisher Chemical Products
500 mL, 500mL
Click to view available options
Quantity:
500 mL
2.5 L
Unit Size:
2.5 L
500mL
Product Code. 10284340
Brand: Fisher Chemical L/2500L/08
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Litmus Solution, Pure, Indicator Grade
TRUSTED_SUSTAINABILITY
Chemical Identifiers
MDL Number 131528
Specifications
Packaging Amber glass bottle
Quantity 500 mL
Physical Form Liquid
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number Certificate Type Date Product Code
Lot Number2531454 Certificate TypeCertificate of Analysis Date24/05/2025 Product CodeL/2500/46,L/2500L/08
Product Identifier
  • Litmus
Product Content Correction
Product Title
Litmus Solution, Pure, Indicator Grade, Fisher Chemical™ >