missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIV-1/ZIP6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14236-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a protein which interacts with the carboxy-terminus of transcription initiation factor TFIIF, a transcription factor which regulates elongation as well as initiation by RNA polymerase II. The protein may also represent a component of an RNA polymerase II holoenzyme complex. Alternative splicing of this gene results in two transcript variants encoding 2 different isoforms.Specifications
LIV-1/ZIP6 | |
Polyclonal | |
Unconjugated | |
SLC39A6 | |
LIV 1, LIV1, SLC39A6, Zinc transporter ZIP6, ZIP 6, ZIP6, Zrt and Irt like protein 6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
106957, 25800, 291733 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q13433, Q4V887, Q8C145 | |
Slc39a6 | |
LIV-1/ZIP6 Fusion Protein Ag5478 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |