missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIV-1/ZIP6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | LIV-1/ZIP6 |
---|---|
Concentration | 0.3 mg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16832784
|
Proteintech
14236-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 13-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16822784
|
Proteintech
14236-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 13-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a protein which interacts with the carboxy-terminus of transcription initiation factor TFIIF, a transcription factor which regulates elongation as well as initiation by RNA polymerase II. The protein may also represent a component of an RNA polymerase II holoenzyme complex. Alternative splicing of this gene results in two transcript variants encoding 2 different isoforms.Specifications
LIV-1/ZIP6 | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Rat, Human, Mouse | |
Q13433, Q4V887, Q8C145 | |
106957, 25800, 291733 | |
LIV-1/ZIP6 Fusion Protein Ag5478 | |
Primary | |
-20°C | |
SLC39A6 |
0.3 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
LIV 1, LIV1, SLC39A6, Zinc transporter ZIP6, ZIP 6, ZIP6, Zrt and Irt like protein 6 | |
Slc39a6 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |