Learn More
LMX1B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 18278-1-AP-150UL
 View more versions of this product
Product Code. 16888344
  • 4652.90 SEK / 150µL
Description
The LIM homeobox transcription factor 1B (LMX1B) belongs to the LIM-homeodomain family. Members of this family are known to be important for pattern formation during development. LMX1B regulates mid-hindbrain patterning; LMX1B-null mice embryos have a severe reduction in the number of midbrain dopaminergic neurons compared to wild-type. While LMX1B appears to be important for both the development and the survival of dopamine neurons, the related LMX1A is crucial for the differentiation of these cells. However, LMX1A and LMX1B function cooperatively to regulated the proliferation, specification and differentiation of midbrain dopaminergic neuronal progenitors.
Specifications
Specifications
LMX1B
Polyclonal
Unconjugated
LMX1B
LIM/homeobox protein 1.2, LIM/homeobox protein LMX1B, LMX 1.2, LMX1.2, LMX1B
Rabbit
Antigen Affinity Chromatography
RUO
114501, 16917, 4010
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Western Blot
0.31 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O60663, O88609
Lmx1b
LMX1B Fusion Protein Ag13096
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Certificates
Promotions
Promotions
Product Content Correction
Product Title
LMX1B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated