Learn More
Lenz Laborglasintrumente Long Ground Sockets
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 01100629
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10539781
  • 845.88 SEK / Pack of 10
Estimated Shipment: 02-07-2024
to see stock.
Add to basket
Description
Description
  • Conical ground sockets
  • Made of DURAN™ tubing
Specifications
Specifications
10/Pk.
2 mm
Borosilicate Glass (3.3)
NS 29/42
33 mm