missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ BioWhittaker™ HEPES Buffer
Used in open vessels incubated under atmospheric conditions. BioWhittaker™ HEPES Buffer is manufactured in accordance with cGMP regulations.
Specifications
Content And Storage | 15° to 30°C |
---|---|
Product Type | HEPES Buffer |
Sterility | Sterile |
pH | 6.98 to 7.3 |
Description
- Used in open vessels incubated under atmospheric conditions
- Behaves as zwitterion
- Sodium chloride concentration of HEPES buffered medium has been reduced to maintain same Osmolality of original media formulations
- Manufactured in accordance with cGMP regulations
- Cell culture-tested to assure compatibility
Specifications
15° to 30°C | |
HEPES Buffer | |
Sterile | |
6.98 to 7.3 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Lonza™ BioWhittaker™ HEPES Buffer