Läs mer
Loperamide hydrochloride, 98+%, Thermo Scientific™
Brand: Thermo Scientific Alfa Aesar J60168.06
| Kvantitet | 5g |
|---|
Se fler versioner av denna produkt
Beskrivning
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- At nanomolar concentrations, binds to mu-opioid receptors
- L-type calcium channel blocker
Kemiska identifierare
| 34552-83-5 | |
| 513.503 | |
| PGYPOBZJRVSMDS-UHFFFAOYSA-N | |
| 71420 | |
| 4-[4-(4-chlorophenyl)-4-hydroxypiperidin-1-yl]-N,N-dimethyl-2,2-diphenylbutanamide;hydrochloride |
| C29H34Cl2N2O2 | |
| MFCD00058581 | |
| loperamide hydrochloride, loperamide hcl, imodium, suprasec, dissenten, fortasec, lopemid, lopemin, loperyl, imosec | |
| CHEBI:6533 | |
| CN(C)C(=O)C(CCN1CCC(CC1)(C2=CC=C(C=C2)Cl)O)(C3=CC=CC=C3)C4=CC=CC=C4.Cl |
Specifikationer
| Loperamide hydrochloride | |
| C29H34Cl2N2O2 | |
| 5g | |
| 14,5571 | |
| PGYPOBZJRVSMDS-UHFFFAOYSA-N | |
| 4-[4-(4-chlorophenyl)-4-hydroxypiperidin-1-yl]-N,N-dimethyl-2,2-diphenylbutanamide;hydrochloride | |
| 71420 | |
| 513.51 |
| 34552-83-5 | |
| MFCD00058581 | |
| UN2811 | |
| loperamide hydrochloride, loperamide hcl, imodium, suprasec, dissenten, fortasec, lopemid, lopemin, loperyl, imosec | |
| CN(C)C(=O)C(CCN1CCC(CC1)(C2=CC=C(C=C2)Cl)O)(C3=CC=CC=C3)C4=CC=CC=C4.Cl | |
| 513.503 | |
| CHEBI:6533 | |
| ≥98% |