Learn More
BRAND™ Soda Lime Glass Culture Tubes with Screw Cap
Manufactured from soda lime glass. BRAND™ Soda Lime Glass Culture Tubes with Screw Cap include a polypropylene screw cap with white elastomer seal and GL thread. Autoclavable at 121°C.
Brand: BRAND™ 113943
Description
- Manufactured from soda lime glass.
- Includes polypropylene screw cap with white elastomer seal and GL thread.
- Autoclavable (121°C).
Specifications
Polypropylene | |
Soda Lime Glass | |
GL 18 | |
Screw Cap |
180 mm | |
100/Pk. | |
18 mm | |
Culture Tube |
Promotions
-
Multiple Manufacturers
Price-Shattering Deals on Lab Glassware
Stock up on lab beakers, flasks and more before these deals disappear.Promo Code: 24123Expires: 2025-06-14