missing translation for 'onlineSavingsMsg'
Learn More

LOXL2 Mouse anti-Human, Unconjugated, Clone: 1H7F1, Proteintech

Mouse Monoclonal Antibody

1656.73 SEK - 4721.67 SEK

Specifications

Antigen LOXL2
Clone 1H7F1
Concentration 1000 ÎĽg/mL
Applications Western Blot, Immunofluorescence, ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16889595
View Documents
Proteintech
67139-1-IG-20UL
20 ÎĽL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16879595
View Documents
Proteintech
67139-1-IG-150UL
150 ÎĽL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

The LOXL2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human LOXL2. This antibody recognizes human antigen. The LOXL2 antibody has been validated for the following applications: IF, WB, ELISA analysis.

Specifications

Specifications

LOXL2
1000 ÎĽg/mL
Monoclonal
Mouse
4017
Primary
1H7F1
Western Blot, Immunofluorescence, ELISA
Unconjugated
Human
IgG2a
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
LOXL2 Mouse anti-Human, Unconjugated, Clone: 1H7F1, Proteintech

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.