Learn More
luciferase Mouse anti-Gaussia Princeps, Unconjugated, Clone: 2D2E1, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67293-1-IG-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16879835
  • 3665.00 SEK / 150µL
Estimated Shipment: 23-03-2023
to see stock.
Description
Description
The luciferase antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of luciferase. This antibody recognizes gaussia princeps antigen. The luciferase antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
luciferase
Monoclonal
1000 μg/mL
Mouse
Primary
IgG2a
Western Blot
2D2E1
Unconjugated
150 μL
Plant