Learn More
luciferase Mouse anti-Gaussia Princeps, Unconjugated, Clone: 2D2E1, Proteintech
Mouse Monoclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | luciferase |
---|---|
Clone | 2D2E1 |
Concentration | 1000 ÎĽg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16889835
|
Proteintech
67293-1-IG-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16879835
|
Proteintech
67293-1-IG-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The luciferase antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of luciferase. This antibody recognizes gaussia princeps antigen. The luciferase antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
luciferase | |
1000 ÎĽg/mL | |
Monoclonal | |
Mouse | |
IgG2a |
2D2E1 | |
Western Blot | |
Unconjugated | |
Plant | |
Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.