Learn More
MP Biomedicals™ Lysing Matrix S, 15 mL Tube
Lysing Matrix S, 15 mL Tube is use to break hard or brittle samples with minimal amounts of mechanical shearing. Specialty applications include RNA and protein extraction from difficult samples where lysis by impaction only is preferable with minimal amounts of mechanical shearing.
Brand: MP Biomedicals™ 116938050
Additional Details : Weight : 0.64000kg
Description
- Matrix Type: S
- Tube Size: 15 mL
- Sample Type: All samples, tough tissues, skeletal muscle, lung, heart, bone, seeds, spores
- Molecule to be Enriched: DNA, RNA, proteins, other small molecules
- Matrix Composition: Matrix S contains stainless steel
Spécification
Stainless Steel | |
Store at Room Temperature. | |
Dnase-free | |
Lysing matrix S | |
RNase-free |
15 mL | |
Bone, Tough Tissues, All Samples, Seeds, Lung, Spores, Skeletal Muscle, Plants, Roots, Plants, Roots, Fixed Samples | |
Nucleic Acid isolation and purification | |
50 x 15 mL | |
Up to 2 g per prep |
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.