Learn More
MP Biomedicals™ Lysing Matrix S, 15 mL Tube
Lysing Matrix S, 15 mL Tube is use to break hard or brittle samples with minimal amounts of mechanical shearing. Specialty applications include RNA and protein extraction from difficult samples where lysis by impaction only is preferable with minimal amounts of mechanical shearing.
651.90 SEK - 4664.00 SEK
Specifications
Capacity | 15 mL |
---|---|
Content And Storage | Store at Room Temperature. |
Sample Type | Bone, Heart, All Samples, Heart, Skeletal Muscle, Seeds, Food, Spores, Food, Plants, Roots, Fixed Samples |
DNase | Dnase-free |
For Use With (Application) | Nucleic Acid isolation and purification |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15272830
|
MP Biomedicals™
116938005 |
5 x 15 mL |
651.90 SEK
Pack of 5 |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15212840
|
MP Biomedicals™
116938050 |
50 x 15 mL |
4664.00 SEK
Pack of 50 |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15282830
|
MP Biomedicals™
116938025 |
25 x 15 mL |
2697.70 SEK
Pack of 25 |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Matrix Type: S
- Tube Size: 15 mL
- Sample Type: All samples, tough tissues, skeletal muscle, lung, heart, bone, seeds, spores
- Molecule to be Enriched: DNA, RNA, proteins, other small molecules
- Matrix Composition: Matrix S contains stainless steel
Specifications
15 mL | |
Bone, Heart, All Samples, Heart, Skeletal Muscle, Seeds, Food, Spores, Food, Plants, Roots, Fixed Samples | |
Nucleic Acid isolation and purification | |
RNase-free |
Store at Room Temperature. | |
Dnase-free | |
Lysing matrix S | |
Up to 2 g per prep |
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.