missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LYZL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17207-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody detects the N-terminal of LYZL1.
The lysozyme family of proteins has an important role in host defense and is associated with the monocyte-macrophage system and enhances the activity of immunoagents. The mammalian lysozyme gene family is much larger than previously appreciated and consists of at least eight distinct genes scattered around the genome. C-type lysozymes are specifically involved catalyzing the hydrolysis of Beta-1, 4 glycosidic bonds of the peptidoglycan of bacterial cell walls. As a homolog of human C-type lysozyme, LYZL1 is an intra-acrosomal, nonbacteriolytic, c lysozyme-like protein recently isolated from human spermatozoa.Specifications
LYZL1 | |
Polyclonal | |
Unconjugated | |
LYZL1 | |
KAAG648, LYC2, lysozyme like 1, Lysozyme like protein 1, LYZL1, PRO1278 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
364745, 67328, 84569 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6UWQ5, Q9CPX3 | |
Lyzl1 | |
LYZL1 Fusion Protein Ag10986 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
LYZL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction