Learn More
MAB21L2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10971-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This antibody is supposed to recognize both MAB21L2 and its homolog MAB21L1.
Required for eye morphogenesis. May promote the survival of proliferating retinal progenitor cells.Specifications
MAB21L2 | |
Polyclonal | |
Unconjugated | |
MAB21L2 | |
mab 21 like 2 (C. elegans), MAB21L2, Protein mab 21 like 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10586, 23937, 680102 | |
-20°C | |
Liquid |
Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8BPP1, Q9Y586 | |
MAB21L2 | |
MAB21L2 Fusion Protein Ag1423 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |