missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAB21L2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | MAB21L2 |
---|---|
Concentration | 0.25 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16827713
|
Proteintech
10971-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 02-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16817713
|
Proteintech
10971-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 02-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This antibody is supposed to recognize both MAB21L2 and its homolog MAB21L1.
Required for eye morphogenesis. May promote the survival of proliferating retinal progenitor cells.Specifications
MAB21L2 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q8BPP1, Q9Y586 | |
10586, 23937, 680102 | |
MAB21L2 Fusion Protein Ag1423 | |
Primary | |
-20°C | |
MAB21L2 |
0.25 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
mab 21 like 2 (C. elegans), MAB21L2, Protein mab 21 like 2 | |
MAB21L2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |