missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAGEA1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12233-1-AP-150UL
This item is not returnable.
View return policy
Description
Human maligt neoplasms carry rejection antigens that are recognized by the patients′ autologous, tumor directed and specific, cytolytic, CD8+ T lymphocyte clones (CTL). An important group of antigens is coded by the MAGE family of genes. It was identified that melanoma associated with antigens (MAAs). Because MAGE-1 is expressed on a significant proportion of human neoplasms of various hisological tyoes (melanoma, brain tumors of glial origin, neuroblastoma, non-small cell lung cancer, breast, gastric, colorectal, ovarian, renal cell carcinomas) and not on normal tissues, the encoded antigen may serve as a marker of early detection and target for cancer immunotherapy.Specifications
MAGEA1 | |
Polyclonal | |
Unconjugated | |
MAGEA1 | |
Antigen MZ2 E, Cancer/testis antigen 1.1, CT1.1, MAGE 1 antigen, MAGE1, MAGE1A, MAGEA1, Melanoma associated antigen 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
4100 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P43355 | |
MAGEA1 | |
MAGEA1 Fusion Protein Ag2871 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MAGEA1 Rabbit anti-Human, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction