Learn More
Invitrogen™ eBioscience™ MagniSort™ Human CD4 Memory T cell Enrichment Kit
Designed for the magnetic separation of CD4 memory T cells by negative selection. It has been optimized for the isolation of human CD4 memory T cells from normal human peripheral blood mononuclear cells utilizing a biotinylated antibody cocktail and streptavidin-coated magnetic beads.
Brand: Invitrogen™ 8804-6813-74
5574.40 SEK valid until 2025-06-30
Use promo code "24074" to get your promotional price.
Alert:
Contains Sodium azide
Includes:
- MagniSort Human CD4 Memory T cell Enrichment Antibody Cocktail : 200 tests, 20ÎĽL/test
- MagniSort Negative Selection Beads A
Description
Undesired cells are bound by antibody and then magnetic beads that, when placed in a magnetic field, leave CD4 memory T cells untouched and free in solution.The MagniSort™ Human CD4 Memory T cell Enrichment Antibody Cocktail contains the following antibodies:
Anti-Human CD8 Biotin
Anti-Human CD11b Biotin
Anti-Human CD14 Biotin
Anti-Human CD16 Biotin
Anti-Human CD19 Biotin
Anti-Human CD20 Biotin
Anti-Human CD36 Biotin
Anti-Human CD45RA Biotin
Anti-Human CD56 Biotin
Anti-Human CD123 Biotin
Anti-Human CD235a Biotin
Anti-Human γδ TCR Biotin
Specifications
2° to 8°C. Do not freeze. | |
Magnetic Cell Separation | |
Aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer | |
200 Tests |