missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mammaglobin A Mouse anti-Human, Clone: 2E5D4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66237-1-IG-20UL
This item is not returnable.
View return policy
Description
This monoclonal antibody is raised against full-length, 93-amino acid human mammaglobin A.
Mammaglobin is a gene that is expressed almost exclusively in the normal breast epithelium and human breast cancer. It is a member of the secretoglobin gene family and forms a heterodimer with lipophilin B. It has been suggested that mammaglobin may be a useful marker for breast cancer clinical research. Studies investigating the detection of mRNA by RT PCR from circulating carcinoma cells in the peripheral blood of breast cancer patients have shown that mammaglobin is a highly specific marker and correlates with several prognostic factors, such as lymph node involvement.Specifications
Mammaglobin A | |
Monoclonal | |
0.87 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q13296 | |
SCGB2A2 | |
Mammaglobin A Fusion Protein Ag22485 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin) | |
2E5D4 | |
Unconjugated | |
SCGB2A2 | |
Mammaglobin 1, Mammaglobin A, MGB1, SCGB2A2, UGB2 | |
Mouse | |
Protein G | |
RUO | |
4250 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Mammaglobin A Mouse anti-Human, Clone: 2E5D4, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction