Learn More
Invitrogen™ MAP3K8 (COT) Recombinant Human Protein
MAP3K8 (COT) is a serine/threonine kinase.
Brand: Invitrogen™ PV4330
Additional Details : Weight : 0.00900kg
Description
MAP3K8 can activate both the MAP and JNK kinase pathways and plays a role in the cell cycle.
Specifications
NP_005195 | |
50mM Tris-HCl pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% Glycerol | |
1mg | |
RUO - research use only | |
MAP3K8 (COT) | |
Human | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
69.9kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Catalytic | |
STE Unique Kinase Family | |
Recombinant | |
Baculovirus |