missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAPK12 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20184-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Activation of members of the mitogen-activated protein kinase family is a major mechanism for transduction of extracellular signals. Stress-activated protein kinases are one subclass of MAP kinases. The protein encoded by this gene functions as a signal transducer during differentiation of myoblasts to myotubes.Specifications
MAPK12 | |
Polyclonal | |
Unconjugated | |
MAPK12 | |
ERK 6, ERK3, ERK6, MAP kinase 12, MAP kinase p38 gamma, MAPK 12, MAPK12, P38GAMMA, PRKM12, SAPK 3, SAPK3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
29857, 60352, 6300 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.42 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O08911, P53778, Q63538 | |
Mapk12 | |
MAPK12 Fusion Protein Ag14073 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |