Learn More
Maspin Mouse anti-Human, Unconjugated, Clone: 3G5A3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67633-1-IG-150UL
Description
The Maspin antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Maspin. This antibody recognizes human antigen. The Maspin antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Maspin | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, Immunohistochemistry, ELISA | |
3G5A3 | |
Unconjugated | |
150 μL | |
5268 | |
IgG2a |