missing translation for 'onlineSavingsMsg'
Learn More

Proteintech MAST1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 13305-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16881444

  • 1355.00 SEK / 20µL
Estimated Shipment: 12-06-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Appears to link the dystrophin/utrophin network with microtubule filaments via the syntrophins. Phosphorylation of DMD or UTRN may modulate their affinities for associated proteins.
Specifications

Specifications

MAST1
Polyclonal
Unconjugated
MAST1
KIAA0973, MAST1, SAST, SAST170
Rabbit
Antigen Affinity Chromatography
RUO
22983, 353118, 56527
-20°C
Liquid
Immunoprecipitation, Western Blot
0.2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q810W7, Q9R1L5, Q9Y2H9
MAST1
MAST1 Fusion Protein Ag4126
20 μL
Primary
Mouse, Rat, Human
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers