Learn More
MAT1A Mouse anti-Human, Unconjugated, Clone: 2F1C1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67408-1-IG-20UL
Description
The MAT1A antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human MAT1A. This antibody recognizes human, mouse, rat antigen. The MAT1A antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
MAT1A | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Western Blot, ELISA | |
2F1C1 | |
Unconjugated | |
20 μL | |
4143 | |
IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.