Learn More
MBL™ Class A Kohlrausch Flask
Expanded neck for easy introduction of viscous liquids or solid samples.
9985.00 SEK - 12280.00 SEK
Specifications
Material | Borosilicate Glass |
---|---|
No. per Pack | 2 |
Product Type | Kohlrausch flask |
Description
Expanded neck for easy introduction of viscous liquids or solid samples.
- Complies with BS 615
- With expanded neck for easy introduction of viscous liquids or solid samples
- 2 sizes available: 100mL for sugar analysis, 200mL for determination of oil viscosity by Engler method
- Calibrated to Class A tolerances
- White enamel inscriptions
- Manufactured from soda-lime glass
Specifications
Borosilicate Glass | |
2 | |
Kohlrausch flask |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MBL™ Class A Kohlrausch Flask