Learn More
MBL™ Class A Kohlrausch Flask
Expanded neck for easy introduction of viscous liquids or solid samples.
9985.00 SEK - 12280.00 SEK
Specifications
Material Borosilicate Glass
No. per Pack 2
Product Type Kohlrausch flask
View More Specs
Products 2
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
15577900
View Documents
MBL
FRE013
200 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15567900
View Documents
MBL
FRE011
100 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
Expanded neck for easy introduction of viscous liquids or solid samples.
  • Complies with BS 615
  • With expanded neck for easy introduction of viscous liquids or solid samples
  • 2 sizes available: 100mL for sugar analysis, 200mL for determination of oil viscosity by Engler method
  • Calibrated to Class A tolerances
  • White enamel inscriptions
  • Manufactured from soda-lime glass
Specifications
Specifications
Borosilicate Glass
2
Kohlrausch flask
Documents
Product Certifications
Product Content Correction
Product Title
MBL™ Class A Kohlrausch Flask