missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCL1L Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15825-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody is specific to MCL1L isoform.
YY1 is a ubiquitously distributed transcription factor belonging to the GLI-Kruppel class of zinc finger proteins. The protein is involved in repressing and activating a diverse number of promoters. YY1 may direct histone deacetylases and histone acetyltransferases to a promoter in order to activate or repress the promoter, thus implicating histone modification in the function of YY1.Specifications
MCL1L | |
Polyclonal | |
Unconjugated | |
MCL1 | |
EAT, Mcl 1, Mcl 1l, Mcl 1s Delta TM, MCL1, mcl1/EAT, MCL1L, MCL1S, TM | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
17210, 4170, 60430 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.34 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P97287, Q07820, Q9Z1P3 | |
Mcl1 | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MCL1L Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction