missing translation for 'onlineSavingsMsg'
Learn More

MCM3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 15597-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16884834

  • 3665.00 SEK / 150µL
Estimated Shipment: 31-03-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene is one of the highly conserved mini-chromosome maintece proteins (MCM) that are involved in the initiation of eukaryotic genome replication. The hexameric protein complex formed by MCM proteins is a key component of the pre-replication complex (pre_RC) and may be involved in the formation of replication forks and in the recruitment of other DNA replication related proteins. This protein is a subunit of the protein complex that consists of MCM2-7. It has been shown to interact directly with MCM5/CDC46. This protein also interacts with, and thus is acetlyated by MCM3AP, a chromatin-associated acetyltransferase. The acetylation of this protein inhibits the initiation of DNA replication and cell cycle progression.
Specifications

Specifications

MCM3
Polyclonal
Unconjugated
MCM3
HCC5, MCM3, P1 MCM3, P1.h, p102, RLF subunit beta, RLFB
Rabbit
Antigen Affinity Chromatography
RUO
17215, 316273, 4172
-20°C
Liquid
Western Blot, Immunoprecipitation
0.18 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P25205, P25206
Mcm3
MCM3 Fusion Protein Ag7966
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers