missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCRS1 Mouse anti-Human, Mouse, Rat, Clone: 2H10E10, Proteintech
Mouse Monoclonal Antibody
1662.46 SEK - 4700.76 SEK
Specifications
Antigen | MCRS1 |
---|---|
Clone | 2H10E10 |
Concentration | 1.5 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16887225
|
Proteintech
60215-1-IG-20UL |
20 ÎĽL |
1662.46 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16877225
|
Proteintech
60215-1-IG-150UL |
150 ÎĽL |
4700.76 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Modulates the transcription repressor activity of DAXX by recruiting it to the nucleolus. As part of the NSL complex it may be involved in acetylation of nucleosomal histone H4 on several lysine residues. Putative regulatory component of the chromatin remodeling INO80 complex which is involved in transcriptional regulation, DNA replication and probably DNA repair. May also be an inhibitor of TERT telomerase activity.Specifications
MCRS1 | |
1.5 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
58 kDa microspherule protein, ICP22BP, INO80 complex subunit J, INO80Q, MCRS1, MCRS2, microspherule protein 1, MSP58, P78 | |
Mcrs1 | |
IgG2a | |
Protein A | |
Antibody |
2H10E10 | |
Western Blot | |
Unconjugated | |
Mouse | |
Mouse, Rat, Human | |
Q96EZ8, Q99L90 | |
10445, 300222, 51812 | |
MCRS1 Fusion Protein Ag1904 | |
Primary | |
-20°C | |
MCRS1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MCRS1 Mouse anti-Human, Mouse, Rat, Clone: 2H10E10, Proteintech