missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEA-1 Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF3088-SP

This item is not returnable.
View return policy
Description
MEA-1 Polyclonal specifically detects MEA-1 in Mouse samples. It is validated for Western Blot.Specifications
MEA-1 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
HYS, male-enhanced antigen, Male-enhanced antigen (H-Y structural gene), male-enhanced antigen 1, MEA1, MEAMEA-1 | |
Goat | |
Affinity Purified | |
RUO | |
4201 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
Q64327 | |
MEA1 | |
E. coli-derived recombinant mouse MEA-1 Ala2-Lys174 Accession # Q64327 | |
25 μg | |
Primary | |
Detects mouse MEA-1 in direct ELISAs and Western blots. | |
Mouse | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
MEA-1 Goat anti-Mouse, Polyclonal, R&D Systems™
Spot an opportunity for improvement?Share a Content Correction