missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MED4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | MED4 |
---|---|
Concentration | 0.31 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16895354
|
Proteintech
15951-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16885354
|
Proteintech
15951-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Mediator is a multiprotein coactivator that is required by DNA-binding transcription factors for activation of polymerase II (see MIM 180660)-transcribed genes. MED4 appears to be a core Mediator subunit and is found in nearly all Mediator preparations (summary by Sato et al., 2004).Specifications
MED4 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Rat, Mouse, Human | |
Q561Q8, Q9CQA5, Q9NPJ6 | |
29079, 306030, 67381 | |
MED4 Fusion Protein Ag8629 | |
Primary | |
-20°C | |
MED4 |
0.31 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
ARC36, DRIP36, HSPC126, MED4, mediator complex subunit 4, RP11 90M2.2, TRAP36, VDRIP | |
MED4 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MED4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech