Learn More
PAL International™ Medipal™ 3 in 1 Disinfectant Wipe Bucket
3.0 L bucket with 350 large disinfectant wipes
Brand: PAL International™ W522110MPCE
This item is not returnable.
View return policy
Description
- Developed in response to a growing clinical need for a single cleaning & disinfectant wipe that is effective against a wider range of pathogens including spores
- It uses a new, high specification combination chemistry, effectively blending two disinfectant compounds, DDAC and Triamine, to produce a highly efficacious, broad spectrum disinfectant effective against spores
- Wipes are alcohol free and intended for cleaning and disinfecting non porous hard surfaces, equipment and non invasive medical devices within healthcare environments
- Proven to kill most micro organisms such as spores, bacteria, viruses & yeasts
- The wipes have been independently tested to a range of European standards (EN) including thenew European test EN16615:2015 (4-field test)
- Alcohol & PHMB Free
- 1 minute contact time
- Heavier weight wipes designed for areas of high use or where additional cleaning and disinfection action is needed
Specifications
Cleans, Disinfects & Sporicidal | |
250 x 275 mm | |
350 wipes in a 3.0 L bucket |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- medipal 3 in 1 disinfectant wipes
Signal Word
- Warning
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : 2-aminoethanol, didecyldimethylammoniumchloride, N-(3-aminopropyl)-N-dodecylpropane-1,3-diamine, al
Product Content Correction
Product Title
PAL International™ Medipal™ 3 in 1 Disinfectant Wipe Bucket > Size: 250 x 275
Spot an opportunity for improvement?Share a Content Correction