missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
MEIS1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-54930
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
MEIS1 Polyclonal specifically detects MEIS1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifikationer
| MEIS1 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 1-4 ug/ml | |
| homeobox protein Meis1, leukemogenic homolog protein, Meis homeobox 1, MGC43380, myeloid ecotropic viral integration site 1 homolog, myeloid ecotropic viral integration site 1 homolog (mouse) | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 4211 | |
| Human | |
| IgG |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| MEIS1 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:GPPLHSHQYPHTAHTNAMAPSMGSSVND | |
| 100 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering