missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEIS2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11550-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a homeobox protein belonging to the TALE (′three amino acid loop extension′) family of homeodomain-containing proteins. TALE homeobox proteins are highly conserved transcription regulators, and several members have been shown to be essential contributors to developmental programs. Multiple transcript variants encoding distinct isoforms have been described for this gene.Specifications
MEIS2 | |
Polyclonal | |
Unconjugated | |
MEIS2 | |
Homeobox protein Meis2, HsT18361, Meis homeobox 2, Meis1 related protein 1, MEIS2, MRG1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
17536, 311311, 4212 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O14770, P97367 | |
Meis2 | |
MEIS2 Fusion Protein Ag2096 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MEIS2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction