missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEP1A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | MEP1A |
---|---|
Concentration | 0.17 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16883125
|
Proteintech
24640-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16873125
|
Proteintech
24640-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MEP1A (Meprin A Subunit Alpha) is a Protein Coding gene. Among its related pathways are Collagen chain trimerization. Gene Ontology (GO) annotations related to this gene include metalloendopeptidase activity and metallopeptidase activity.Specifications
MEP1A | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Rat, Mouse | |
P28825, Q16819, Q64230 | |
17287, 25684, 4224 | |
MEP1A Fusion Protein Ag18950 | |
Primary | |
-20°C | |
MEP1A |
0.17 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Endopeptidase 2, MEP1A, Meprin A subunit alpha, PABA peptide hydrolase, PPH alpha, PPHA | |
Mep1a | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MEP1A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech