Learn More
Mercury Standard Metal Solution 10,000ppm, for ICP, Solution HNO3 1M, Fisher Chemical Product Code.: 10785331
Mercury Standard Metal Solution 10,000ppm, for ICP, Solution HNO3 1M, Fisher Chemical
Product Code. 10785331 Shop All Fisher Chemical Products
100 mL
Click to view available options
Quantity:
100 mL
Product Code. 10785331
Brand: Fisher Chemical J/8292/05
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Packaging HDPE plastic bottle
Physical Form Liquid
MDL Number 11035
Color Colorless
pH 1
Quantity 100 mL
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Mercury solution 10 000 ppm in ca. M nitric acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H290-May be corrosive to metals.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Nitric acid, Mercury
  • REA17-CMR1-Restricted to professional users
Product Content Correction
Product Title
Mercury Standard Metal Solution 10,000ppm, for ICP, Solution HNO3 1M, Fisher Chemical >