missing translation for 'onlineSavingsMsg'
Learn More

Methyl Violet in Acetic Acid 99 to 100% Indicator Solution For Laboratory Use and Indicator For Nonaqueous Titrations, Chem Lab

Brand:  Chem Lab CL07.1303.0100


Product Code. 11997433

  • 594.00 SEK / 100mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

Purple
17°C
Glass bottles
40 °C
118°C
1.05 g/mL
100 mL
Liquid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Methyl violet in acetic acid 99-100% indicator solution

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H400-Very toxic to aquatic life.

Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : acetic acid

SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methyl Violet in Acetic Acid 99 to 100% Indicator Solution For Laboratory Use and Indicator For Nonaqueous Titrations, Chem Lab > Methyl Violet in Acetic Acid 99 to 100% Indicator Solution For Laboratory Use and Indicator For Nonaqueous Titrations, 100 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.