missing translation for 'onlineSavingsMsg'
Learn More

SP Bel-Art™ Metric Color Octagon Spinbar Magnetic Stirring Bars

Interrupted profile provides greater surface area and added turbulence when compared to the smooth surface of cylindrical bars

Brand:  SP Bel-Art™ F37108-1303

Additional Details : Weight : 0.00570kg

 View more versions of this product

Product Code. 16805383

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • The most commonly used shape.
  • Choose from red, yellow or blue.
  • Pivot ring allows the Spin Bar to “rock” while spinning for better mixing conditions
Specifications

Specifications

3 mm
No Pivot Ring
10 Pk.
Autoclavable
PTFE, FDA-grade, USP Class VI
Yellow
PTFE
13 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers