missing translation for 'onlineSavingsMsg'
Learn More

SP Bel-Art™ Metric Color Octagon Spinbar Magnetic Stirring Bars

Interrupted profile provides greater surface area and added turbulence when compared to the smooth surface of cylindrical bars

Brand:  SP Bel-Art™ F37108-5108

Additional Details : Weight : 0.04540kg

 View more versions of this product

Product Code. 16865383

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • The most commonly used shape.
  • Choose from red, yellow or blue.
  • Pivot ring allows the Spin Bar to “rock” while spinning for better mixing conditions
Specifications

Specifications

8 mm
Yellow
PTFE
51 mm
PTFE, FDA-grade, USP Class VI
5 Pk.
Autoclavable
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers