missing translation for 'onlineSavingsMsg'
Learn More

SP Bel-Art™ Metric Color Octagon Spinbar Magnetic Stirring Bars

Interrupted profile provides greater surface area and added turbulence when compared to the smooth surface of cylindrical bars

Brand:  SP Bel-Art™ F37108-7613

Additional Details : Weight : 0.03970kg

 View more versions of this product

Product Code. 16875383

  • 150.00 SEK / Each
Estimated Shipment: 02-07-2024
to see stock.
Add to basket

Description

Description

  • The most commonly used shape.
  • Choose from red, yellow or blue.
  • Pivot ring allows the Spin Bar to “rock” while spinning for better mixing conditions
Specifications

Specifications

13 mm
Yellow
Autoclavable
PTFE, FDA-grade, USP Class VI
PTFE
76 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers