Learn More
ROBU VitraPOR™ Micro-Filter Candle, Cylindrical Shape
Micro-Filter Candle without glass tube.
Brand: ROBU VitraPOR™ 18001
Additional Details : Weight : 0.02140kg
Description
- Highly resistant to most chemicals and temperatures up to 500°C
Specifications
Borosilicate Glass (3.3) | |
Non-sterile | |
15 mm |
1 μm | |
1 | |
6 mm |