Learn More
ROBU VitraPOR™ Micro-Filter Candle, Cylindrical Shape
Micro-Filter Candle without glass tube.
1570.00 SEK - 4730.00 SEK
Specifications
Material Borosilicate Glass 3.3
Sterility Non-sterile
View More Specs
Products 26
Product Code Brand Diameter (Metric) Length (Metric) Porosity Price Quantity & Availability  
Product Code Brand Diameter (Metric) Length (Metric) Porosity Price Quantity & Availability  
16872926
View Documents Promotion Details
ROBU VitraPOR™
18044
20 mm 55 mm 4
3890.00 SEK
Pack of 5
View alternative products
Save Up to 
»
Estimated Shipment: 16-06-2023
to see stock.
Add to basket
16852906
View Documents Promotion Details
ROBU VitraPOR™
18005
6 mm 15 mm 5
3235.00 SEK
Pack of 5
View alternative products
Save Up to 
»
Estimated Shipment: 16-06-2023
to see stock.
Add to basket
16892906
View Documents Promotion Details
ROBU VitraPOR™
18013
9 mm 20 mm 3
1595.00 SEK
Pack of 5
View alternative products
Save Up to 
»
Estimated Shipment: 16-06-2023
to see stock.
Add to basket
16892916
View Documents Promotion Details
ROBU VitraPOR™
18031
20 mm 30 mm 1
2665.00 SEK
Pack of 5
View alternative products
Save Up to 
»
Estimated Shipment: 16-06-2023
to see stock.
Add to basket
16842916
View Documents Promotion Details
ROBU VitraPOR™
18022
13 mm 25 mm 2
1625.00 SEK
Pack of 5
View alternative products
Save Up to 
»
Estimated Shipment: 16-06-2023
to see stock.
Add to basket
16802906
View Documents Promotion Details
ROBU VitraPOR™
18000
6 mm 15 mm 0
1570.00 SEK
Pack of 5
View alternative products
Save Up to 
»
Estimated Shipment: 16-06-2023
to see stock.
Add to basket
16802916
View Documents Promotion Details
ROBU VitraPOR™
18014
9 mm 20 mm 4
1780.00 SEK
Pack of 5
View alternative products
Save Up to 
»
Estimated Shipment: 16-06-2023
to see stock.
Add to basket
16812916
View Documents Promotion Details
ROBU VitraPOR™
18015
9 mm 20 mm 5
3870.00 SEK
Pack of 5
View alternative products
Save Up to 
»
Estimated Shipment: 16-06-2023
to see stock.
Add to basket
16822926
View Documents Promotion Details
ROBU VitraPOR™
18034
20 mm 30 mm 4
3065.00 SEK
Pack of 5
View alternative products
Save Up to 
»
Estimated Shipment: 16-06-2023
to see stock.
Add to basket
16832906
View Documents Promotion Details
ROBU VitraPOR™
18003
6 mm 15 mm 3
1570.00 SEK
Pack of 5
View alternative products
Save Up to 
»
Estimated Shipment: 16-06-2023
to see stock.
Add to basket
16822916
View Documents Promotion Details
ROBU VitraPOR™
18020
13 mm 25 mm 0
1625.00 SEK
Pack of 5
View alternative products
Save Up to 
»
Estimated Shipment: 16-06-2023
to see stock.
Add to basket
16862926
View Documents Promotion Details
ROBU VitraPOR™
18043
20 mm 55 mm 3
3315.00 SEK
Pack of 5
View alternative products
Save Up to 
»
Estimated Shipment: 16-06-2023
to see stock.
Add to basket
16882916
View Documents Promotion Details
ROBU VitraPOR™
18030
20 mm 30 mm 0
2665.00 SEK
Pack of 5
View alternative products
Save Up to 
»
Estimated Shipment: 16-06-2023
to see stock.
Add to basket
16832926
View Documents Promotion Details
ROBU VitraPOR™
18040
20 mm 55 mm 0
3315.00 SEK
Pack of 5
View alternative products
Save Up to 
»
Estimated Shipment: 16-06-2023
to see stock.
Add to basket
16812906
View Documents Promotion Details
ROBU VitraPOR™
18001
6 mm 15 mm 1
1570.00 SEK
Pack of 5
View alternative products
Save Up to 
»
Estimated Shipment: 16-06-2023
to see stock.
Add to basket
16862916
View Documents Promotion Details
ROBU VitraPOR™
18024
13 mm 25 mm 4
1820.00 SEK
Pack of 5
View alternative products
Save Up to 
»
Estimated Shipment: 16-06-2023
to see stock.
Add to basket
16842926
View Documents Promotion Details
ROBU VitraPOR™
18041
20 mm 55 mm 1
3315.00 SEK
Pack of 5
View alternative products
Save Up to 
»
Estimated Shipment: 16-06-2023
to see stock.
Add to basket
16802926
View Documents Promotion Details
ROBU VitraPOR™
18032
20 mm 30 mm 2
2665.00 SEK
Pack of 5
View alternative products
Save Up to 
»
Estimated Shipment: 16-06-2023
to see stock.
Add to basket
16812926
View Documents Promotion Details
ROBU VitraPOR™
18033
20 mm 30 mm 3
2665.00 SEK
Pack of 5
View alternative products
Save Up to 
»
Estimated Shipment: 16-06-2023
to see stock.
Add to basket
16842906
View Documents Promotion Details
ROBU VitraPOR™
18004
6 mm 15 mm 4
1745.00 SEK
Pack of 5
View alternative products
Save Up to 
»
Estimated Shipment: 16-06-2023
to see stock.
Add to basket
16872916
View Documents Promotion Details
ROBU VitraPOR™
18025
13 mm 25 mm 5
4730.00 SEK
Pack of 5
View alternative products
Save Up to 
»
Estimated Shipment: 16-06-2023
to see stock.
Add to basket
16862906
View Documents Promotion Details
ROBU VitraPOR™
18010
9 mm 20 mm 0
1595.00 SEK
Pack of 5
View alternative products
Save Up to 
»
Estimated Shipment: 16-06-2023
to see stock.
Add to basket
16852916
View Documents Promotion Details
ROBU VitraPOR™
18023
13 mm 25 mm 3
1625.00 SEK
Pack of 5
View alternative products
Save Up to 
»
Estimated Shipment: 16-06-2023
to see stock.
Add to basket
16872906
View Documents Promotion Details
ROBU VitraPOR™
18011
9 mm 20 mm 1
1595.00 SEK
Pack of 5
View alternative products
Save Up to 
»
Estimated Shipment: 16-06-2023
to see stock.
Add to basket
16852926
View Documents Promotion Details
ROBU VitraPOR™
18042
20 mm 55 mm 2
3315.00 SEK
Pack of 5
View alternative products
Save Up to 
»
Estimated Shipment: 16-06-2023
to see stock.
Add to basket
16832916
View Documents Promotion Details
ROBU VitraPOR™
18021
13 mm 25 mm 1
1625.00 SEK
Pack of 5
View alternative products
Save Up to 
»
Estimated Shipment: 16-06-2023
to see stock.
Add to basket
Description
Description
  • Highly resistant to most chemicals and temperatures up to 500°C
Specifications
Specifications
Borosilicate Glass 3.3
Non-sterile