Learn More
ROBU VitraPOR™ Micro-Filter Candle, Cylindrical Shape
Micro-Filter Candle without glass tube.
1570.00 SEK - 4730.00 SEK
Product Code | Brand | Diameter (Metric) | Length (Metric) | Porosity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Length (Metric) | Porosity | Price | Quantity & Availability | |||
16872926
|
ROBU VitraPOR™
18044 |
20 mm | 55 mm | 4 |
3890.00 SEK
Pack of 5 |
Log in to see stock. |
16852906
|
ROBU VitraPOR™
18005 |
6 mm | 15 mm | 5 |
3235.00 SEK
Pack of 5 |
Log in to see stock. |
16892906
|
ROBU VitraPOR™
18013 |
9 mm | 20 mm | 3 |
1595.00 SEK
Pack of 5 |
Log in to see stock. |
16892916
|
ROBU VitraPOR™
18031 |
20 mm | 30 mm | 1 |
2665.00 SEK
Pack of 5 |
Log in to see stock. |
16842916
|
ROBU VitraPOR™
18022 |
13 mm | 25 mm | 2 |
1625.00 SEK
Pack of 5 |
Log in to see stock. |
16802906
|
ROBU VitraPOR™
18000 |
6 mm | 15 mm | 0 |
1570.00 SEK
Pack of 5 |
Log in to see stock. |
16802916
|
ROBU VitraPOR™
18014 |
9 mm | 20 mm | 4 |
1780.00 SEK
Pack of 5 |
Log in to see stock. |
16812916
|
ROBU VitraPOR™
18015 |
9 mm | 20 mm | 5 |
3870.00 SEK
Pack of 5 |
Log in to see stock. |
16822926
|
ROBU VitraPOR™
18034 |
20 mm | 30 mm | 4 |
3065.00 SEK
Pack of 5 |
Log in to see stock. |
16832906
|
ROBU VitraPOR™
18003 |
6 mm | 15 mm | 3 |
1570.00 SEK
Pack of 5 |
Log in to see stock. |
16822916
|
ROBU VitraPOR™
18020 |
13 mm | 25 mm | 0 |
1625.00 SEK
Pack of 5 |
Log in to see stock. |
16862926
|
ROBU VitraPOR™
18043 |
20 mm | 55 mm | 3 |
3315.00 SEK
Pack of 5 |
Log in to see stock. |
16882916
|
ROBU VitraPOR™
18030 |
20 mm | 30 mm | 0 |
2665.00 SEK
Pack of 5 |
Log in to see stock. |
16832926
|
ROBU VitraPOR™
18040 |
20 mm | 55 mm | 0 |
3315.00 SEK
Pack of 5 |
Log in to see stock. |
16812906
|
ROBU VitraPOR™
18001 |
6 mm | 15 mm | 1 |
1570.00 SEK
Pack of 5 |
Log in to see stock. |
16862916
|
ROBU VitraPOR™
18024 |
13 mm | 25 mm | 4 |
1820.00 SEK
Pack of 5 |
Log in to see stock. |
16842926
|
ROBU VitraPOR™
18041 |
20 mm | 55 mm | 1 |
3315.00 SEK
Pack of 5 |
Log in to see stock. |
16802926
|
ROBU VitraPOR™
18032 |
20 mm | 30 mm | 2 |
2665.00 SEK
Pack of 5 |
Log in to see stock. |
16812926
|
ROBU VitraPOR™
18033 |
20 mm | 30 mm | 3 |
2665.00 SEK
Pack of 5 |
Log in to see stock. |
16842906
|
ROBU VitraPOR™
18004 |
6 mm | 15 mm | 4 |
1745.00 SEK
Pack of 5 |
Log in to see stock. |
16872916
|
ROBU VitraPOR™
18025 |
13 mm | 25 mm | 5 |
4730.00 SEK
Pack of 5 |
Log in to see stock. |
16862906
|
ROBU VitraPOR™
18010 |
9 mm | 20 mm | 0 |
1595.00 SEK
Pack of 5 |
Log in to see stock. |
16852916
|
ROBU VitraPOR™
18023 |
13 mm | 25 mm | 3 |
1625.00 SEK
Pack of 5 |
Log in to see stock. |
16872906
|
ROBU VitraPOR™
18011 |
9 mm | 20 mm | 1 |
1595.00 SEK
Pack of 5 |
Log in to see stock. |
16852926
|
ROBU VitraPOR™
18042 |
20 mm | 55 mm | 2 |
3315.00 SEK
Pack of 5 |
Log in to see stock. |
16832916
|
ROBU VitraPOR™
18021 |
13 mm | 25 mm | 1 |
1625.00 SEK
Pack of 5 |
Log in to see stock. |
Description
- Highly resistant to most chemicals and temperatures up to 500°C
Specifications
Borosilicate Glass 3.3 | |
Non-sterile |