missing translation for 'onlineSavingsMsg'
Learn More

ROBU VitraPOR™ Micro-Filter Candle, Cylindrical Shape

Micro-Filter Candle without glass tube.

Brand:  ROBU VitraPOR™ 18023

Additional Details : Weight : 0.03470kg

 View more versions of this product

Product Code. 16852916

  • 1820.00 SEK / Pack of 5
Estimated Shipment: 12-06-2024
to see stock.
Add to basket

Description

Description

  • Highly resistant to most chemicals and temperatures up to 500°C
Specifications

Specifications

Borosilicate Glass (3.3)
3
13 mm
Non-sterile
25 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers