Learn More
ROBU VitraPOR™ Micro-Filter Candle, Cylindrical Shape
Micro-Filter Candle without glass tube.
Brand:  ROBU VitraPOR™ 18011
Additional Details : Weight : 0.02520kg
 View more versions of this product
Product Code. 16872906
  • 1785.00 SEK / Pack of 5
Estimated Shipment: 24-06-2024
to see stock.
Add to basket
Description
Description
  • Highly resistant to most chemicals and temperatures up to 500°C
Specifications
Specifications
Borosilicate Glass (3.3)
Non-sterile
20 mm
1 μm
1
9 mm