missing translation for 'onlineSavingsMsg'
Learn More

ROBU VitraPOR™ Micro-Filter Candle, Cylindrical Shape

Micro-Filter Candle without glass tube.

Brand:  ROBU VitraPOR™ 18030

Additional Details : Weight : 0.07410kg

 View more versions of this product

Product Code. 16882916

  • 2665.00 SEK / Pack of 5

Estimated Shipment: 20-06-2023
to see stock.
Add to basket

Description

Description

  • Highly resistant to most chemicals and temperatures up to 500°C
Specifications

Specifications

Borosilicate Glass 3.3
0
20 mm
Non-sterile
30 mm
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers