Learn More
ROBU VitraPOR™ Micro-Filter Candle with Glass Tube
Micro-Filter Candle with narrow glass tube.
Brand: ROBU VitraPOR™ 18115
Additional Details : Weight : 0.10120kg
Description
- Highly resistant to most chemicals and temperatures up to 500°C
Specifications
Borosilicate Glass 3.3 | |
Glass Tube | |
20 mm |
Non-sterile | |
5 | |
9 mm |