missing translation for 'onlineSavingsMsg'
Learn More

Robu™ Micro-Filter-Candles

Robu™ Micro-Filter-Candles are highly resistant to most chemicals such as solvents, acids, and alkalis.

Brand:  Robu™ 18133

Additional Details : Weight : 0.26700kg

 View more versions of this product

Product Code. 16882946

  • 3450.00 SEK / Pack of 5

Estimated Shipment: 17-04-2023
to see stock.
Add to basket

Description

Description

  • ISO 4793-80
  • Cylindrical shape
  • With narrow glass tube
Specifications

Specifications

Cylindrical
Narrow glass tube
180 mm
20 mm
16 to 40 μm
Filter Candle
30 mm
20 mm
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers